7-Fluoro-1H-benzo[d]imidazole-2-carbaldehyde 97%

Supplier: Ambeed
Warning

A312603-100MG A312603-50MG
77194-678EA 272.95 USD
77194-678 77194-680
7-Fluoro-1H-benzo[d]imidazole-2-carbaldehyde 97%
7-Fluoro-1H-benzo[d]imidazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅FN₂O
MW: 164.13 g/mol
MDL Number: MFCD18807799
CAS Number: 615568-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR