4-(Cyclopropylcarbamoyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Cyclopropylcarbamoyl)benzeneboronic acid, 4-(Cyclopropylaminocarbonyl)phenylboronic acid

A311559-10G A311559-1G A311559-250MG A311559-5G
77167-120EA 204.97 USD
77167-120 77167-122 77167-124 77167-126
4-(Cyclopropylcarbamoyl)phenylboronic acid 97%
4-(Cyclopropylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂BNO₃
MW: 205.02 g/mol
MDL Number: MFCD03788419
CAS Number: 515140-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR