2-Methoxy-4-methyl-5-pyridineboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-Methoxy-4-methylpyridin-3-ylboronic acid, 2-Methoxy-4-methylpyridine-5-boronic acid

A311297-250MG A311297-100MG A311297-1G A311297-5G
77167-116EA 39 USD
77167-116 77892-637 77892-638 77892-639
2-Methoxy-4-methyl-5-pyridineboronic acid 95%
2-Methoxy-4-methyl-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₃
MDL Number: MFCD07368880
CAS Number: 503184-35-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR