Zinc(II) Bis(trifluoromethanesulfonyl)imide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Zinc bis(trifluoromethylsulphonyl)imide, Bis(trifluoromethanesulphonyl)imide Zinc(II) Salt, Zinc(II) Triflimide
A311037-1G
A311037-250MG
A311037-25G
A311037-5G
77167-108EA
12
USD
77167-108
77167-110
77167-112
77167-114
Zinc(II) Bis(trifluoromethanesulfonyl)imide 98%
Zinc(II) Bis(trifluoromethanesulfonyl)imide
Precaución: for Research Use Only
Formula:
C₄F₁₂N₂O₈S₄Zn MW: 625.65 g/mol |
MDL Number:
MFCD16621474 CAS Number: 168106-25-0 UN: 1759 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...