5-(Difluoromethyl)-1H-pyrazole-5-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A310559-100MG
A310559-1G
A310559-250MG
77194-604EA
41.2
USD
77194-604
77194-606
77194-608
5-(Difluoromethyl)-1H-pyrazole-5-carboxylic acid 97%
5-(Difluoromethyl)-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₄F₂N₂O₂ MW: 162.094 g/mol |
MDL Number:
MFCD18809604 CAS Number: 681034-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...