4-(Methylthio)benzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Methylthio)benzyl Alcohol

A310330-100G A310330-1G A310330-25G A310330-5G
77194-596EA 461 USD
77194-596 77194-598 77194-600 77194-602
4-(Methylthio)benzyl alcohol 98%
4-(Methylthio)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₀OS
MDL Number: MFCD00009706
CAS Number: 3446-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR