1-(5-Bromo-3-pyridyl)piperazine 96%

Supplier: Ambeed
Warning

A310287-100MG A310287-5G
77194-592EA 25 USD
77194-592 77194-594
1-(5-Bromo-3-pyridyl)piperazine 96%
1-(5-Bromo-3-pyridyl)piperazine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN₃
MDL Number: MFCD09910230
CAS Number: 412347-30-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR