Pyrrolidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A309819-1G A309819-250MG A309819-25G A309819-5G
77167-094EA 21 USD
77167-094 77167-096 77167-098 77167-100
Pyrrolidine-3-carboxylic acid 95%
Pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C5H9NO2
MDL Number: MFCD01318699
CAS Number: 59378-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR