tert-Butyl (5-(trifluoromethyl)pyridin-3-yl)carbamate 97%

Supplier: Ambeed
Warning

A309653-25G A309653-5G
77167-086EA 135.96 USD
77167-086 77167-088
tert-Butyl (5-(trifluoromethyl)pyridin-3-yl)carbamate 97%
tert-Butyl (5-(trifluoromethyl)pyridin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₃N₂O₂
MW: 262.23 g/mol
MDL Number: MFCD22201173
CAS Number: 1187055-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR