5-Cyclopropylpyrazin-2-amine 95%

Supplier: Ambeed
Warning

A309515-100MG A309515-1G A309515-250MG
77167-080EA 149.35 USD
77167-080 77593-858 77683-914
5-Cyclopropylpyrazin-2-amine 95%
5-Cyclopropylpyrazin-2-amine

Precaución: For research use only.

Formula: C₇H₉N₃
MW: 135.17 g/mol
MDL Number: MFCD12032675
CAS Number: 920313-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR