2-Amino-5-nitropyrimidin-4(1H)-one 95%

Supplier: Ambeed
Warning

A309131-100MG A309131-1G A309131-250MG
77167-074EA 68 USD
77167-074 77167-076 77167-078
2-Amino-5-nitropyrimidin-4(1H)-one 95%
2-Amino-5-nitropyrimidin-4(1H)-one

Precaución: for Research Use Only

Formula: C₄H₄N₄O₃
MDL Number: MFCD22495111
CAS Number: 7254-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR