(2-(1-(3,5-Bis(trifluoromethyl)benzyl)-5-(pyridin-4-yl)-1H-1,2,3-triazol-4-yl)pyridin-3-yl)(2-chlorophenyl)methanone 99%

Supplier: Ambeed
Warning

Synonyms: Tradipitant

A308950-50MG A308950-100MG
77194-578EA 200.85 USD
77194-578 77550-858
(2-(1-(3,5-Bis(trifluoromethyl)benzyl)-5-(pyridin-4-yl)-1H-1,2,3-triazol-4-yl)pyridin-3-yl)(2-chlorophenyl)methanone 99%
(2-(1-(3,5-Bis(trifluoromethyl)benzyl)-5-(pyridin-4-yl)-1H-1,2,3-triazol-4-yl)pyridin-3-yl)(2-chlorophenyl)methanone

Precaución: for Research Use Only

Formula: C₂₈H₁₆ClF₆N₅O
MW: 587.9 g/mol
MDL Number: MFCD28127604
CAS Number: 622370-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR