1-(Phenylsulfonyl)-3-indoleboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Phenylsulphonyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A308684-250MG A308684-5G
77194-564EA 9 USD
77194-564 77571-858
1-(Phenylsulfonyl)-3-indoleboronic acid pinacol ester 98%
1-(Phenylsulphonyl)-3-indoleboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₂BNO₄S
MDL Number: MFCD08276778
CAS Number: 870717-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR