Methyl 5-amino-2-bromo-4-methylbenzoate 97%

Supplier: Ambeed
Warning

A308556-10G A308556-1G A308556-250MG A308556-25G A308556-5G
77167-066EA 260.59 USD
77167-066 77167-068 77167-070 77167-072 77612-776
Methyl 5-amino-2-bromo-4-methylbenzoate 97%
Methyl 5-amino-2-bromo-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.08 g/mol
MDL Number: MFCD14706185
CAS Number: 474330-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR