Pyridine-2-aldoxime methochloride 98%

Supplier: Ambeed
Danger

Synonyms: Pyridine-2-carboxaldoxime methochloride, Pralidoxime Chloride, 2-PAM chloride, 2-((Hydroxyimino)methyl)-1-methylpyridin-1-ium chloride

A307244-100G A307244-100MG A307244-10G A307244-25G A307244-50MG
77194-532EA 120.51 USD
77194-532 77194-534 77194-536 77194-538 77194-540
Pyridine-2-aldoxime methochloride 98%
Pyridine-2-aldoxime methochloride

Precaución: for Research Use Only

Formula: C₇H₉ClN₂O
MW: 172.61 g/mol
MDL Number: MFCD00011981
CAS Number: 51-15-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR