1-Cyclopentylurea 95+%

Supplier: Ambeed
Warning

A306615-1G A306615-250MG A306615-5G
77167-056EA 33 USD
77167-056 77167-058 77167-060
1-Cyclopentylurea 95+%
1-Cyclopentylurea

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O
MDL Number: MFCD00019276
CAS Number: 1194-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR