2-Bromo-3-fluoropyridin-4-amine 95%

Supplier: Ambeed
Warning

A306549-1G A306549-250MG A306549-50MG
77167-050EA 416.12 USD
77167-050 77167-052 77167-054
2-Bromo-3-fluoropyridin-4-amine 95%
2-Bromo-3-fluoropyridin-4-amine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
MW: 191.00 g/mol
MDL Number: MFCD28679452
CAS Number: 1417638-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR