Decernotinib (VX-509) 98+%

Supplier: Ambeed
Warning

Synonyms: (R,Z)-2-((2-(1H-pyrrolo[2,3-b]pyridin-3-yl)pyrimidin-4-yl)amino)-2-methyl-N-(2,2,2-trifluoroethyl)butanimidic acid, (R)-2-((2-(1H-Pyrrolo[2,3-b]pyridin-3-yl)pyrimidin-4-yl)amino)-2-methyl-N-(2,2,2-trifluoroethyl)butanamide, Decernotinib

A306535-10MG A306535-25MG A306535-2MG A306535-5MG A306535-1MG A306535-50MG
77167-042EA 56.65 USD
77167-042 77167-044 77167-046 77167-048 77612-760 77683-907
Decernotinib (VX-509) 98+%
Decernotinib (VX-509)

Precaución: For research use only.

Formula: C₁₈H₁₉F₃N₆O
MW: 392.38 g/mol
MDL Number: MFCD27987896
CAS Number: 944842-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR