1,8-Dihydroxy-9,10-anthraquinone 98%

Supplier: Ambeed
Warning

Synonyms: Dantron, Danthron, 1,8-Dihydroxyanthraquinone

A306488-10G A306488-1MG A306488-100G A306488-25G A306488-500G
77194-530EA 17.51 USD
77194-530 77616-324 77746-584 77746-585 77746-586
1,8-Dihydroxy-9,10-anthraquinone 98%
1,8-Dihydroxy-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₈O₄
MW: 240.22 g/mol
MDL Number: MFCD00001211
CAS Number: 117-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR