Diethyl 2-aminothiophene-3,4-dicarboxylate 98%
Supplier: Ambeed
|
Warning
|
A306442-1G
A306442-250MG
A306442-5G
77167-036EA
30
USD
77167-036
77167-038
77167-040
Diethyl 2-aminothiophene-3,4-dicarboxylate 98%
Diethyl 2-aminothiophene-3,4-dicarboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₄S |
MDL Number:
MFCD08059079 CAS Number: 104680-25-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...