3-(Aminomethyl)-4-methylpyridin-2(1H)-one hydrochloride 97%

Supplier: Ambeed
Warning

A305445-100MG A305445-1G A305445-250MG
77194-504EA 96 USD
77194-504 77194-506 77194-508
3-(Aminomethyl)-4-methylpyridin-2(1H)-one hydrochloride 97%
3-(Aminomethyl)-4-methylpyridin-2(1H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂O
MDL Number: MFCD25541769
CAS Number: 2301857-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR