5-Chloro-2-methylphenylhydrazine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-2-hydrazinotoluene hydrochloride, (5-Chloro-2-methylphenyl)hydrazine hydrochloride

A305264-1G A305264-5G
77194-500EA 10.3 USD
77194-500 77194-502
5-Chloro-2-methylphenylhydrazine hydrochloride 97%
5-Chloro-2-methylphenylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClN₂·ClH
MW: 193.074 g/mol
MDL Number: MFCD04038837
CAS Number: 5446-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR