2,3-Difluoro-6-methoxyphenol 95+%

Supplier: Ambeed
Danger

A304654-100MG A304654-1G A304654-250MG
77167-014EA 30 USD
77167-014 77892-619 77892-620
2,3-Difluoro-6-methoxyphenol 95+%
2,3-Difluoro-6-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆F₂O₂
MDL Number: MFCD04116024
CAS Number: 186306-70-7
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR