tert-Butyl 4-(1-hydroxyethyl)piperidine-1-carboxylate 96%

Supplier: Ambeed
Warning

A304500-1G A304500-250MG A304500-5G
77167-008EA 98 USD
77167-008 77167-010 77167-012
tert-Butyl 4-(1-hydroxyethyl)piperidine-1-carboxylate 96%
tert-Butyl 4-(1-hydroxyethyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₃
MDL Number: MFCD15474944
CAS Number: 183170-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR