tert-Butyl 3-amino-3-(hydroxymethyl)azetidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A304329-100MG A304329-250MG
77167-004EA 156.56 USD
77167-004 77167-006
tert-Butyl 3-amino-3-(hydroxymethyl)azetidine-1-carboxylate 95%
tert-Butyl 3-amino-3-(hydroxymethyl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₃
MW: 202.25 g/mol
MDL Number: MFCD18375152
CAS Number: 1262411-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR