6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinolin-1(2H)-one 95%

Supplier: Ambeed
Warning

A304307-100MG A304307-1G A304307-250MG A304307-5G
77166-996EA 37 USD
77166-996 77166-998 77167-000 77167-002
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinolin-1(2H)-one 95%
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₅H₁₈BNO₃
MDL Number: MFCD18426474
CAS Number: 1219130-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR