5-(4-Chlorophenyl)-1-methyl-1H-pyrrole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A303565-100MG A303565-1G A303565-250MG
77194-488EA 55.62 USD
77194-488 77194-490 77194-492
5-(4-Chlorophenyl)-1-methyl-1H-pyrrole-2-carboxylic acid 95%
5-(4-Chlorophenyl)-1-methyl-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO₂
MW: 235.67 g/mol
MDL Number: MFCD10006412
CAS Number: 1017414-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR