2-Fluoro-3-isopropoxybenzoic acid 97%

Supplier: Ambeed
Warning

A302955-1G A302955-250MG
77194-478EA 71.07 USD
77194-478 77746-574
2-Fluoro-3-isopropoxybenzoic acid 97%
2-Fluoro-3-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.19 g/mol
MDL Number: MFCD28356761
CAS Number: 1346608-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR