(S)-2-Amino-2-(3-fluorophenyl)ethanol 95%

Supplier: Ambeed
Warning

A302718-1G A302718-5G A302718-100MG A302718-250MG
77166-984EA 84 USD
77166-984 77166-986 77892-613 77892-614
(S)-2-Amino-2-(3-fluorophenyl)ethanol 95%
(S)-2-Amino-2-(3-fluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₀FNO
MDL Number: MFCD09256370
CAS Number: 325152-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR