1,3-Bis(4-chlorophenyl)imidazolium chloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Bis(4-chlorophenyl)-1H-imidazol-3-ium chloride
A302024-100MG
A302024-1G
A302024-250MG
A302024-5G
77194-468EA
25.75
USD
77194-468
77194-470
77194-472
77746-573
1,3-Bis(4-chlorophenyl)imidazolium chloride 98%
1,3-Bis(4-chlorophenyl)imidazolium chloride
Precaución: for Research Use Only
Formula:
C₁₅H₁₁Cl₃N₂ MW: 325.620 g/mol |
MDL Number:
MFCD03788924 CAS Number: 141556-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...