2-Cyclohexylacetonitrile 97%

Supplier: Ambeed
Danger

A300702-10G A300702-1G A300702-250MG A300702-25G
77166-970EA 318 USD
77166-970 77166-972 77166-974 77166-976
2-Cyclohexylacetonitrile 97%
2-Cyclohexylacetonitrile

Precaución: for Research Use Only

Formula: C₈H₁₃N
MW: 123.196 g/mol
MDL Number: MFCD00058667
CAS Number: 4435-14-7
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR