3-Hydroxy-4-methoxypyridine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-4-methoxypicolinic acid

A299939-100MG A299939-1G A299939-250MG
77194-442EA 96.82 USD
77194-442 77194-444 77194-446
3-Hydroxy-4-methoxypyridine-2-carboxylic acid 97%
3-Hydroxy-4-methoxypyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₄
MW: 169.135 g/mol
MDL Number: MFCD18254272
CAS Number: 210300-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR