cis-3-Aminocyclobutanecarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A298966-100MG
A298966-1G
A298966-250MG
A298966-5G
77166-938EA
36
USD
77166-938
77166-940
77166-942
77166-944
cis-3-Aminocyclobutanecarboxylic acid 97%
cis-3-Aminocyclobutanecarboxylic acid
Precaución: for Research Use Only
Formula:
C₅H₉NO₂ |
MDL Number:
MFCD06659964 CAS Number: 74316-27-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...