3-Fluoro-5-iodopyridine 98%

Supplier: Ambeed
Warning

A298906-100MG A298906-1G A298906-250MG A298906-25G A298906-5G A298906-100G
77166-928EA 12.36 USD
77166-928 77166-930 77166-932 77166-934 77166-936 77683-891
3-Fluoro-5-iodopyridine 98%
3-Fluoro-5-iodopyridine

Precaución: For research use only.

Formula: C₅H₃FIN
MDL Number: MFCD08457780
CAS Number: 757950-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR