7-Methoxyisobenzofuran-1(3H)-one 97%

Supplier: Ambeed
Warning

A298634-250MG A298634-100MG
77194-432EA 64 USD
77194-432 77550-810
7-Methoxyisobenzofuran-1(3H)-one 97%
7-Methoxyisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₉H₈O₃
MDL Number: MFCD15146118
CAS Number: 28281-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR