2-Amino-4-chlorothiazole-5-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A298514-100MG
A298514-10G
A298514-1G
A298514-250MG
A298514-5G
77166-920EA
9.27
USD
77166-920
77166-922
77166-924
77166-926
77683-889
2-Amino-4-chlorothiazole-5-carbaldehyde 95%
2-Amino-4-chlorothiazole-5-carbaldehyde
Precaución: For research use only.
Formula:
C₄H₃ClN₂OS MW: 162.5974 g/mol |
MDL Number:
MFCD00204993 CAS Number: 76874-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...