3-Amino-6-chloroisonicotinamide 97%

Supplier: Ambeed
Warning

A298318-100MG A298318-250MG A298318-1G
77194-422EA 72 USD
77194-422 77194-424 77616-296
3-Amino-6-chloroisonicotinamide 97%
3-Amino-6-chloroisonicotinamide

Precaución: for Research Use Only

Formula: C₆H₆ClN₃O
MDL Number: MFCD12026342
CAS Number: 1217026-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR