5-Chloro-2-nitropyridin-3-ol 97%

Supplier: Ambeed
Warning

A297665-1G A297665-250MG
77194-412EA 215.27 USD
77194-412 77194-414
5-Chloro-2-nitropyridin-3-ol 97%
5-Chloro-2-nitropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
MW: 174.54 g/mol
MDL Number: MFCD18259798
CAS Number: 936247-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR