Oxiran-2-ylmethyl 4-methylbenzenesulfonate 97%

Supplier: Ambeed
Warning

A295910-1G A295910-5G
77166-912EA 33.99 USD
77166-912 77166-914
Oxiran-2-ylmethyl 4-methylbenzenesulfonate 97%
Oxiran-2-ylmethyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄S
MW: 228.26 g/mol
MDL Number: MFCD00066850
CAS Number: 6746-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR