2-Cyano-3-methoxypyridine 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 3-Methoxypicolinonitrile, 3-methoxypyridine-2-carbonitrile

A295829-100MG A295829-10G A295829-1G A295829-5G A295829-250MG
77166-904EA 44.29 USD
77166-904 77166-906 77166-908 77166-910 77683-882
2-Cyano-3-methoxypyridine 95%, Ambeed.Inc
2-Cyano-3-methoxypyridine

Precaución: For research use only.

Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD07374875
CAS Number: 24059-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR