6-Bromo-5-iodopyridin-2-amine 95%

Supplier: Ambeed
Warning

A294970-100MG A294970-1G A294970-250MG A294970-5G
77166-880EA 28.84 USD
77166-880 77166-882 77166-884 77166-886
6-Bromo-5-iodopyridin-2-amine 95%
6-Bromo-5-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrIN₂
MW: 298.90 g/mol
MDL Number: MFCD22570450
CAS Number: 1806995-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR