Boc-(4-aminomethylphenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Boc-aminomethylphenylacetic acid, 2-(4-(((tert-Butoxycarbonyl)amino)methyl)phenyl)acetic acid

A294746-1G A294746-250MG A294746-5G A294746-100MG
77166-876EA 57 USD
77166-876 77166-878 77593-796 77892-598
Boc-(4-aminomethylphenyl)acetic acid 98%
Boc-(4-aminomethylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MDL Number: MFCD06656436
CAS Number: 71420-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR