5-(4-Bromophenyl)-4,6-dichloropyrimidine 97%

Supplier: Ambeed
Warning

A294502-10G A294502-1G A294502-25G A294502-5G
77166-858EA 41 USD
77166-858 77166-860 77166-862 77166-864
5-(4-Bromophenyl)-4,6-dichloropyrimidine 97%
5-(4-Bromophenyl)-4,6-dichloropyrimidine

Precaución: for Research Use Only

Formula: C₁₀H₅BrCl₂N₂
MDL Number: MFCD13151897
CAS Number: 146533-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR