1,4-Diformylpiperazine 98%

Supplier: Ambeed
Warning

A294409-25G A294409-5G
77166-854EA 47.38 USD
77166-854 77166-856
1,4-Diformylpiperazine 98%
1,4-Diformylpiperazine

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O₂
MW: 142.16 g/mol
MDL Number: MFCD00006153
CAS Number: 4164-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR