2-(2-Cyano-4-methoxyphenyl)acetic acid 98%

Supplier: Ambeed
Warning

A294062-100MG A294062-250MG
77166-848EA 59.74 USD
77166-848 77166-850
2-(2-Cyano-4-methoxyphenyl)acetic acid 98%
2-(2-Cyano-4-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MW: 191.183 g/mol
MDL Number: MFCD18394271
CAS Number: 52786-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR