Methyl 1-methyl-1,2,3,6-tetrahydropyridine-4-carboxylate 95%

Supplier: Ambeed
Warning

A293441-100MG A293441-1G A293441-250MG
77166-828EA 65 USD
77166-828 77166-830 77166-832
Methyl 1-methyl-1,2,3,6-tetrahydropyridine-4-carboxylate 95%
Methyl 1-methyl-1,2,3,6-tetrahydropyridine-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₃NO₂
MDL Number: MFCD05237238
CAS Number: 59097-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR