Methyl 4-methyl-5-nitropicolinate 97%

Supplier: Ambeed
Warning

A293078-1G A293078-250MG
77166-822EA 33 USD
77166-822 77892-589
Methyl 4-methyl-5-nitropicolinate 97%
Methyl 4-methyl-5-nitropicolinate

Precaución: for Research Use Only

Formula: C₈H₈N₂O₄
MDL Number: MFCD22421724
CAS Number: 868551-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR