1,2,3,6-Tetrahydropyridine hydrochloride 97%

Supplier: Ambeed
Warning

A292469-250MG A292469-5G A292469-1G
77194-368EA 9.27 USD
77194-368 77194-370 77616-272
1,2,3,6-Tetrahydropyridine hydrochloride 97%
1,2,3,6-Tetrahydropyridine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClN
MW: 119.6 g/mol
MDL Number: MFCD00216642
CAS Number: 18513-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR