3-Chloro-5,6-dimethylpyrazin-2(1H)-one 97%

Supplier: Ambeed
Warning

A292310-1G A292310-250MG
77194-364EA 152 USD
77194-364 77194-366
3-Chloro-5,6-dimethylpyrazin-2(1H)-one 97%
3-Chloro-5,6-dimethylpyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MDL Number: MFCD28957066
CAS Number: 140862-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR