4-(2-Hydroxyethyl)pyridine 95%

Supplier: Ambeed
Warning

A292233-1G A292233-250MG A292233-5G
77194-358EA 34 USD
77194-358 77194-360 77194-362
4-(2-Hydroxyethyl)pyridine 95%
4-(2-Hydroxyethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD00129038
CAS Number: 5344-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR